missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUBA1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583674
This item is not returnable.
View return policy
Description
Immunogen sequence: ATYAPVISAE KAYHEQLSVA EITN Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.
Specifications
| TUBA1A | |
| Polyclonal | |
| Unconjugated | |
| TUBA1A | |
| alpha 1a antibody; alpha 3 antibody; alpha-tubulin; alpha-tubulin 1; Alpha-tubulin 3; alpha-tubulin 3 antibody; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; B-ALPHA-1 antibody; brain-specific antibody; Detyrosinated tubulin alpha-1A chain; FLJ25113 antibody; FLJ30169; H2 ALPHA; H2ALPHA; H2-ALPHA; hum-a-tub1; hum-a-tub1 antibody; hum-a-tub2; hum-a-tub2 antibody; LIS3; LIS3 antibody; LOC106557476; M40; MGC117247; MGC16435; OK/SW cl.56; OK/SWcl.56; OK/SW-cl.56; Talpha1 alpha-tubulin; TUBA1; Tuba-1; Tuba1a; TUBA1A antibody; TUBA3; TUBA3 antibody; TUBA6; TUBB1; TUBB5; tubulin alpha 1a; tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-1A chain antibody; tubulin alpha-3 chain; tubulin alpha-3 chain antibody; Tubulin B-alpha-1; tubulin B-alpha-1 antibody; tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 1A; tubulin, alpha 6; tubulin, alpha, brain-specific; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22142, 64158, 7846 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P68369, P68370, Q71U36 | |
| TUBA1A | |
| Recombinant protein corresponding to Human TUBA1A. Recombinant protein control fragment (Product #RP-104359). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction