missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody (RM113), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77401
This item is not returnable.
View return policy
Description
alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in All samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP)
Specifications
| alpha Tubulin | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| All species | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
| RM113 | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1:200, Immunoprecipitation 1:200, Chromatin Immunoprecipitation (ChIP) 1:200 | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| This alpha Tubulin Antibody (RM113) was developed against a peptide corresponding to the C-terminus of alpha Tubulin | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| 7846 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction