missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody (DM1AR), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-38646SS
This item is not returnable.
View return policy
Description
alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in nan samples. It is validated for nan
Specifications
| alpha Tubulin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:5000, Flow Cytometry 1 μg per million cells, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1 μg/mL, Immunohistochemistry-Paraffin 1:100-1:500, Flow (Intracellular) 1 μg per million cells | |
| alpha tubulin loading control, alpha tubulin monoclonal, Alpha-tubulin 3, B-ALPHA-1, DM1A, DM1A anti-tubulin, dm1a monoclonal, DM1a tubulin, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA1 monoclonal, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin monoclonal, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| This Recombinant alpha Tubulin Antibody (DM1AR) was developed against native chicken brain microtubules. | |
| 0.025 mg | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| 7846 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry | |
| DM1AR | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Not Applicable | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction