missing translation for 'onlineSavingsMsg'
Learn More

alpha Tubulin 4a Rabbit anti-Human, Mouse, Rat, Clone: 1K6U1, Novus Biologicals™

Product Code. 18339544 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339544 100 μg 100µL
18339026 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18339544 Supplier Novus Biologicals Supplier No. NBP316646100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

alpha Tubulin 4a Monoclonal antibody specifically detects alpha Tubulin 4a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Tubulin 4a
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1K6U1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:20-1:50
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias alpha 1 (testis specific), Alpha-tubulin 1, FLJ30169, H2-ALPHA, Testis-specific alpha-tubulin, TUBA1, Tubulin alpha-1 chain, tubulin alpha-4A chain, Tubulin H2-alpha, tubulin, alpha 1, tubulin, alpha 4a
Host Species Rabbit
Immunogen A synthesized peptide derived from human alpha Tubulin 4a
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7277
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.