missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha TTP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528781
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32, mouse liver, rat liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a soluble protein that binds alpha-trocopherol, a form of vitamin E, with high selectivity and affinity. This protein plays an important role in regulating vitamin E levels in the body by transporting vitamin E between membrane vesicles and facilitating the secretion of vitamin E from hepatocytes to circulating lipoproteins. Mutations in this gene cause hereditary vitamin E deficiency (ataxia with vitamin E deficiency, AVED) and retinitis pigmentosa.
Specifications
| alpha TTP | |
| Polyclonal | |
| Unconjugated | |
| Ttpa | |
| alpha tocopherol transfer protein; alpha TTP; alpha-tocopherol transfer protein; alphaTTP; alpha-TTP; ATTP; AVED; tocopherol (alpha) transfer protein; tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin E deficiency); Tocopherol transfer protein alpha; TPP1; TTP; TTP1; TTPA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100751868, 25571, 50500, 7274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P41034, P49638, Q8BWP5 | |
| Ttpa | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human alpha TTP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Hamster, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction