missing translation for 'onlineSavingsMsg'
Learn More

Alpha-TTP Antibody (7B5), Novus Biologicals™

Product Code. 18382438 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382438 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382438 Supplier Novus Biologicals Supplier No. H00007274M0150ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Alpha-TTP Monoclonal antibody specifically detects Alpha-TTP in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha-TTP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7B5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000361.1
Gene Alias alpha-tocopherol transfer protein, alphaTTP, Alpha-TTP, ataxia (Friedreich-like) with vitamin E deficiency, ATTP, AVED, tocopherol (alpha) transfer protein, tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin Edeficiency), TPP1, TTP1
Host Species Mouse
Immunogen TTPA (NP_000361.1, 179 a.a. ~ 278 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IAAVLTDSFPLKVRGIHLINEPVIFHAVFSMIKPFLTEKIKERIHMHGNNYKQSLLQHFPDILPLEYGGEEFSMEDICQEWTNFIMKSEDYLSSISESIQ
Purification Method Protein A or G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7274
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.