missing translation for 'onlineSavingsMsg'
Learn More

alpha-Toluenesulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog Number 11449372
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1939-99-7
C7H7ClO2S
190.641
MFCD00007455
OAHKWDDSKCRNFE-UHFFFAOYSA-N
alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride
74740
phenylmethanesulfonyl chloride
C1=CC=C(C=C1)CS(=O)(=O)Cl
This item is not returnable. View return policy
1939-99-7
C7H7ClO2S
190.641
MFCD00007455
OAHKWDDSKCRNFE-UHFFFAOYSA-N
alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride
74740
phenylmethanesulfonyl chloride
C1=CC=C(C=C1)CS(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 89°C to 94°C
Boiling Point 120°C (10 mmHg)
Quantity 5 g
UN Number UN3096
Beilstein 972806
Sensitivity Moisture sensitive
Formula Weight 190.65
Percent Purity 99%
Chemical Name or Material alpha-Toluenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'