missing translation for 'onlineSavingsMsg'
Learn More

alpha-Toluenesulfonyl chloride, 99%

Product Code. 11469372
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469372 100 g 100g
11449372 5 g 5g
11459372 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469372 Supplier Thermo Scientific Alfa Aesar Supplier No. A10124.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1939-99-7 | C7H7ClO2S | 190.641 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1939-99-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00007455
InChI Key OAHKWDDSKCRNFE-UHFFFAOYSA-N
Synonym alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride
PubChem CID 74740
IUPAC Name phenylmethanesulfonyl chloride
SMILES C1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Melting Point 89°C to 94°C
Boiling Point 120°C (10 mmHg)
Quantity 100 g
UN Number UN3096
Beilstein 972806
Sensitivity Moisture sensitive
Formula Weight 190.65
Percent Purity 99%
Chemical Name or Material alpha-Toluenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.