missing translation for 'onlineSavingsMsg'
Learn More

alpha-Tocopheryl Succinate, TRC

Product Code. 30135166
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135166 5 g 5g
30135141 10 g 10g
30009583 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135166 Supplier Toronto Research Chemicals Supplier No. TRCT5399505G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4345-03-3
Molecular Formula C33 H54 O5
Molecular Weight (g/mol) 530.78
Synonym RRR-alpha-Tocopheryl hydrogen succinate, (2R)-2,5,7,8-Tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl hydrogen succinate, D-alpha-Tocopheryl Hydrogen Succinate, Butanedioic acid, 1-[(2R)-3,4-dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-yl] ester (ACI), 6-Chromanol, 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-, hydrogen succinate, (+)- (8CI), Butanedioic acid, mono[(2R)-3,4-dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-yl] ester (9CI), Butanedioic acid, mono[3,4-dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-1-benzopyran-6-yl] ester, [2R-[2R*(4R*,8R*)]]- (ZCI), Succinic acid, mono[2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-6-chromanyl] ester, (+)- (8CI), α-Tocopherol, succinate (6CI), (+)-α-Tocopheryl succinate, Covitol 1210, CV 104, d-α-Tocopherol acid succinate, d-α-Tocopheryl acid succinate, d-α-Tocopheryl hemisuccinate, NSC 173849, Tocopherol succinate, Vitamin E acid succinate, Vitamin E d-α-tocosuccinate, Vitamin E hemisuccinate, Vitamin E succinate, α-Tocopherol acid succinate, α-Tocopherol hemisuccinate, α-Tocopheryl acid succinate, α-Tocopheryl hemisuccinate, α-Tocopheryl succinate, α-TOS, α-Vitamin E succinate
IUPAC Name 4-oxo-4-[[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl]oxy]butanoic acid
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)CCC(=O)O)c(C)c(C)c2O1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C33H54O5/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-33(8)21-19-28-27(7)31(25(5)26(6)32(28)38-33)37-30(36)18-17-29(34)35/h22-24H,9-21H2,1-8H3,(H,34,35)/t23-,24-,33-/m1/s1
Formula Weight 530.3971
Chemical Name or Material RRR-alpha-Tocopheryl hydrogen succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.