missing translation for 'onlineSavingsMsg'
Learn More

(+)-alpha-Tocopherol Acid Succinate, Mikromol

Product Code. 30458974
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30458974 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458974 Supplier Mikromol Supplier No. MM0167.030250
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(+)-alpha-Tocopherol Acid Succinate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material RRR-alpha-Tocopheryl hydrogen succinate
CAS 4345-03-3
Analyte or Component Names RRR-alpha-Tocopheryl hydrogen succinate
Shipping Condition Room Temperature
Molecular Formula C33 H54 O5
Certifications/Compliance ISO 17034
Synonym RRR-alpha-Tocopheryl hydrogen succinate, (2R)-2,5,7,8-Tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl hydrogen succinate, d-alpha-tocopherol succinate, Vitamin E succinate, (+)-alpha-tocopheryl acid succinate, D-alpha-Tocopheryl hydrogen succinate
InChI Formula InChI=1S/C33H54O5/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-33(8)21-19-28-27(7)31(25(5)26(6)32(28)38-33)37-30(36)18-17-29(34)35/h22-24H,9-21H2,1-8H3,(H,34,35)/t23-,24-,33-/m1/s1
Solution Type Neat
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)CCC(=O)O)c(C)c(C)c2O1
IUPAC Name 4-oxo-4-[[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl]oxy]butanoic acid
Molecular Weight (g/mol) 530.78
Quantity 250 mg
Formula Weight 530.3971 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.