missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Tocopherol Acetate, Alpha-Tocopherol Acetate, TRC

Product Code. 30008252
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008252 25 g 25g
17907824 1 g 1g
17979154 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008252 Supplier Toronto Research Chemicals Supplier No. TRCT52611525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C31 H52 O3
Molecular Weight (g/mol) 472.74
Synonym (+)-alpha-Tocopheryl Acetate, RRR-alpha-Tocopheryl acetate, (+)-alpha-Tocopheryl Acetate, (2R)-3,4-dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-Benzopyran-6-ol 6-acetate, 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-, acetate, [2R-[2R*(4R*,8R*)]]-, 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-, acetate, (2R)- (9CI), 6-Chromanol, 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-, acetate, (+)- (8CI), Vitamin E acetate (7CI), α-Tocopherol acetate (6CI), (+)-α-Tocopherol acetate, (+)-α-Tocopheryl acetate, (2R,4'R,8'R)-α-Tocopherol acetate, (2R,4'R,8'R)-α-Tocopheryl acetate, (R,R,R)-α-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)-6-chromanol acetate, Alfacol, Combinal E, Contopheron, Copherol 12250, Copherol 1250, Copherol F 1250C, Covitol, Covitol 1100, Covitol 1360, Covitol 700, Covitol 700WD, D-α-Tocopherol acetate, D-α-Tocopheryl acetate, E 230 Clear, E-Ferol, E-Toplex, E-Vicotrat, Ecofrol, Econ, Endo E Dompe, Ephynal acetate, Epsilan-M, Erevit, Evicup, Evipherol, Fertilvit, Gevex, Lutavit E 50, Nanotopes, NatAc, Natur-E granulate, Spondyvit, T-3376, Tinoderm E, Tocopherex, Tocopherol acetate, Tocopheryl acetate, Tocophrin, Tofaxin, Tokoferol acetate, Vitamin E 230 Clear, Vitamin Eα acetate, Vitapherol, d-Vitamin E acetate, d-α-Tocopherol acetate, d-α-Tocopheryl acetate, α-Tocopheryl acetate
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)C)c(C)c(C)c2O1

Specifications

CAS 58-95-7
Quantity 25 g
InChI Formula InChI=1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1
Formula Weight 472.3916
Percent Purity >95%
Chemical Name or Material RRR-alpha-Tocopheryl acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.