missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpineol, 96%, Thermo Scientific Chemicals

Catalog Number p-7049619
15.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
98-55-5
C10H18O
154.253
MFCD00001557
WUOACPNHFRMFPN-UHFFFAOYSA-N
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
17100
CHEBI:22469
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
CC1=CCC(CC1)C(C)(C)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
98-55-5
C10H18O
154.253
MFCD00001557
WUOACPNHFRMFPN-UHFFFAOYSA-N
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
17100
CHEBI:22469
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
CC1=CCC(CC1)C(C)(C)O

A naturally-occuring monoterpene alcohol

A naturally-occuring monoterpene alcohol.Alpha-terpineol is used as an antioxidant, antiseptic, antihypernociception and anti-inflammatory. It is also used as a solvent. It is an important ingredient of pine oil disinfectants. Further, it is used as a fragrance in perfumes, fat denaturant for soap production and synthetic flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 31°C to 32°C
Color Colorless to Yellow
Density 0.930 g/mL
Boiling Point 210°C to 219°C
Flash Point 90°C (194°F)
Refractive Index 1.483
Quantity 25 g
Merck Index 14,9171
Solubility Information Miscible with water,benzene,propylene glycol,acetone,alcohol and ether.
Formula Weight 154.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material alpha-Terpineol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'