missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpinene, 90+%

Product Code. p-7042542
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 15485169

Brand: Thermo Scientific Alfa Aesar J62871.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-86-5 | C10H16 | 136.24 g/mol

It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.

Solubility
Soluble in ether, alcohol, and ethanol (100 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: -20°C. Store under inert gas. Air sensitive. Keep away from strong acids, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001534
InChI Key YHQGMYUVUMAZJR-UHFFFAOYSA-N
Synonym alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
PubChem CID 7462
ChEBI CHEBI:10334
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,3-diene
SMILES CC(C)C1=CC=C(C)CC1

Specifications

Color Colorless
Density 0.837
Boiling Point 173°C to 175°C
Flash Point 50°C (122°F)
Refractive Index 1.478
Quantity 100 mL
UN Number UN2319
Beilstein 1853379
Merck Index 14,9170
Solubility Information Soluble in ether,alcohol,and ethanol (100mg/ml). Insoluble in water.
Formula Weight 136.23
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material alpha-Terpinene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • alpha-Terpinene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : alpha-Terpinene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.