missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ alpha-Synuclein Antibody (prasinezumab) - Humanized, Novus Biologicals™

Human Monoclonal Antibody

£157.00 - £695.00

Specifications

Antigen alpha-Synuclein
Clone prasinezumab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30044761
View Documents
Novus Biologicals™
NBP3-28363-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30044160
View Documents
Novus Biologicals™
NBP3-28363-1mg
1 mg
£695.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30047943
View Documents
Novus Biologicals™
NBP3-28363-100ug
100 μg
£256.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

alpha-Synuclein
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein
Alpha-synuclein
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
prasinezumab
Monoclonal
Purified
RUO
Human
Q9Y6H5
6622
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.