missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody (5C2), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04321AF350
This item is not returnable.
View return policy
Description
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| alpha-Synuclein | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SNCA | |
| This alpha-Synuclein Antibody (5C2) was developed against recombinant human alpha-synuclein (61-95) purified from E. coli (NP_0003360). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5C2 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| Mouse | |
| Protein G purified | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction