missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alpha-Smooth Muscle Actin Recombinant Rabbit Monoclonal Antibody (23GB600), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554092
This item is not returnable.
View return policy
Description
Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown that smooth muscle actin appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases. Multiple alternatively spliced variants of smooth muscle actin have been identified.
Specifications
| Alpha-Smooth Muscle Actin | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62736, P62737, P62738 | |
| ACTA2 | |
| A synthetic peptide of human alpha smooth muscle Actin (1-54AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 23GB600 | |
| Unconjugated | |
| ACTA2 | |
| 0610041G09Rik; AAT6; ACT-4; acta2; acta2.L; acta2.S; actin; Actin Alpha 2 Smooth Muscle Aorta; actin alpha 2, smooth muscle; Actin Vascular Smooth Muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha 2, smooth muscle, aorta L homeolog; actin, alpha 2, smooth muscle, aorta S homeolog; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; ACTSA; ACTVS; alpha actin 2; Alpha Cardiac Actin; alpha smooth muscle actin; alpha-actin; Alpha-actin-2; alpha-cardiac actin; alphaSMA; alpha-SM-actin; alpha-smooth muscle actin; Aortic Smooth Muscle; asma; a-sma; cell growth-inhibiting gene 46 protein; GIG46; Growth Inhibiting Gene 46; hypothetical protein LOC515610; LOW QUALITY PROTEIN: actin, aortic smooth muscle; mymy5; SM actin; sma; SMactin; sm-actin; SMalphaA; smooth muscle; smooth muscle alpha-actin; vascular smooth muscle alpha-actin; wu:fb63d03; XELAEV_18034370mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11475, 59, 81633 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction