missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alpha-Smooth Muscle Actin Polyclonal Antibody
GREENER_CHOICE

Product Code. 11356263
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356263 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11356263 Supplier Invitrogen™ Supplier No. PA518292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown that smooth muscle actin appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases. Multiple alternatively spliced variants of smooth muscle actin have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha-Smooth Muscle Actin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ACTA2
Gene Accession No. P62736, P62737, P62738
Gene Alias 0610041G09Rik; AAT6; ACT-4; acta2; acta2.L; acta2.S; actin; Actin Alpha 2 Smooth Muscle Aorta; actin alpha 2, smooth muscle; Actin Vascular Smooth Muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha 2, smooth muscle, aorta L homeolog; actin, alpha 2, smooth muscle, aorta S homeolog; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; ACTSA; ACTVS; alpha actin 2; Alpha Cardiac Actin; alpha smooth muscle actin; alpha-actin; Alpha-actin-2; alpha-cardiac actin; alphaSMA; alpha-SM-actin; alpha-smooth muscle actin; Aortic Smooth Muscle; asma; a-sma; cell growth-inhibiting gene 46 protein; GIG46; Growth Inhibiting Gene 46; hypothetical protein LOC515610; LOW QUALITY PROTEIN: actin, aortic smooth muscle; mymy5; SM actin; sma; SMactin; sm-actin; SMalphaA; smooth muscle; smooth muscle alpha-actin; vascular smooth muscle alpha-actin; wu:fb63d03; XELAEV_18034370mg
Gene Symbols ACTA2
Host Species Goat
Immunogen Synthetic peptide sequence (EEEDSTALVC) corresponding to the N-terminus amino acids of ACTA2 (aa 3-12).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11475, 59, 81633
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.