missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Mouse anti-Human, Clone: 132D3, Novus Biologicals™
Mouse Monoclonal Antibody
£259.00 - £407.00
Specifications
| Antigen | alpha-Smooth Muscle Actin |
|---|---|
| Clone | 132D3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18258232
|
Novus Biologicals
NBP2-59429-50UL |
50 μL |
£259.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18251971
|
Novus Biologicals
NBP2-59429-100UL |
100 μL |
£407.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| alpha-Smooth Muscle Actin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Stem Cell Markers | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 59 | |
| Recombinant human ACTA2 (3-377aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 132D3 | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin | |
| ACTA2 | |
| IgG1 κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title