missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 28 publications
£220.00
Specifications
| Antigen | alpha-Smooth Muscle Actin |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.3 - 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA Detection limit 1:32000, Immunohistochemistry Free-Floating |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
alpha-Smooth Muscle Actin Polyclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Free-Floating.Specifications
| alpha-Smooth Muscle Actin | |
| Western Blot 0.3 - 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA Detection limit 1:32000, Immunohistochemistry Free-Floating | |
| Polyclonal | |
| Goat | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin | |
| ACTA2 | |
| IgG | |
| Affinity Purified | |
| Variants NP_001604.1 and NP_001135417.1 encode the same protein. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P62736 | |
| 59 | |
| Alpha-Smooth Muscle Actin Antibody is made to a peptide with sequence EEEDSTALVC corresponding to N-Terminus according to NP_001604.1, NP_001135417.1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title