missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Antibody (1A4/asm-1) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
alpha-Smooth Muscle Actin Monoclonal antibody specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Baboon, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF
Specifications
Specifications
| Antigen | alpha-Smooth Muscle Actin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), CyTOF |
| Classification | Monoclonal |
| Clone | 1A4/asm-1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Simple Western, Flow Cytometry, ELISA 1-5ug/mL for coating, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/mL, Immunohistochemistry-Frozen 0.5-1ug/mL, Flow (Intracellular), CyTOF-ready |
| Formulation | PBS |
| Gene Alias | AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?