missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alpha Sarcoglycan Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106600
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SGCA.
The sarcoglycan transmembrane proteins are members of the dystrophin complex. Sarcoglycans cluster together to form a complex, which is localized in the cell membrane of skeletal, cardiac, and smooth muscle fibers. Four sarcoglycan subunit proteins, designated alpha-, beta-, gamma- and delta-sarcoglycan, form a complex on the skeletal muscle cell surface membrane. A genetic defect in any one of these proteins causes the loss or marked decrease of the whole sarcoglycan complex, which is observed in the autosomal recessive muscular dystrophy, sarcoglycanopathy. In smooth muscle, beta- and delta-sarcoglycans are associated with epsilon-sarcoglycan, a glycoprotein homologous to alpha-sarcoglycan. Additionally, a complete deficiency in delta-sarcoglycan is the cause of the Syrian hamster BIO.14 cardiomyopathy.
Specifications
| Alpha Sarcoglycan | |
| Polyclonal | |
| Unconjugated | |
| SGCA | |
| 50 kDa dystrophin-associated glycoprotein; 50DAG; 50kD DAG; adhalin; ADL; Alpha-sarcoglycan; alpha-SG; Asg; DAG2; DMDA2; Dystroglycan 2; Dystroglycan2; dystroglycan-2; LGMD2D; limb girdle muscular dystrophy 2D; sarcoglycan alpha; sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein); sarcoglycan, alpha (dystrophin-associated glycoprotein); SCARMD1; SG alpha; SG-alpha; Sgca | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20391, 303468, 6442 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P82350, Q16586 | |
| SGCA | |
| A synthesized peptide derived from human SGCA(Accession Q16586), corresponding to amino acid residues P167-D217. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction