missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Phenylcyclopentylacetic acid, 98%
Click to view available options
Quantity:
5 g
Unit Size:
5g
Description
alpha-Phenylcyclopentylacetic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
alpha-Phenylcyclopentylacetic acid is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
alpha-Phenylcyclopentylacetic acid is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Specifications
Specifications
| Melting Point | 98°C to 102°C |
| Quantity | 5 g |
| Beilstein | 1956835 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 204.27 |
| Percent Purity | 98% |
| Chemical Name or Material | alpha-Phenylcyclopentylacetic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction