missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Phenyl-2-pyridineacetamide, TRC

Product Code. 30428543
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428543 5 g -
30420140 250 mg 250mg
30413660 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428543 Supplier Toronto Research Chemicals Supplier No. TRCP3365305G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7251-52-7
Molecular Formula C13 H12 N2 O
Molecular Weight (g/mol) 212.25
Synonym 2-Pyridineacetamide, α-phenyl-, 2-Phenyl-2-(2-pyridyl)acetamide, NSC 62572, SC 16571, α-(2-Pyridyl)benzeneacetamide, (2RS)-2-Phenyl-2-(pyridin-2-yl)acetamide, Methylphenidate Hydrochloride Imp. F (EP)
IUPAC Name 2-phenyl-2-pyridin-2-ylacetamide
SMILES NC(=O)C(c1ccccc1)c2ccccn2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H12N2O/c14-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-15-11/h1-9,12H,(H2,14,16)
Formula Weight 212.095
Chemical Name or Material a-Phenyl-2-pyridineacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.