missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-parvin/Actopaxin Antibody (OTI1C10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72018APC
This item is not returnable.
View return policy
Description
Alpha-parvin/Actopaxin Monoclonal antibody specifically detects Alpha-parvin/Actopaxin in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Alpha-parvin/Actopaxin | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI1C10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| actopaxin, alpha-parvin, Calponin-like integrin-linked kinase-binding protein, CH-ILKBP, FLJ10793, FLJ12254, FLJ25106, FLJ46049, Matrix-remodeling-associated protein 2, matrix-remodelling associated 2, MXRA2, parvin, alpha | |
| Full length human recombinant protein of human PARVA(NP_060692) produced in HEK293T cell. | |
| 0.1 mL | |
| Phospho Specific | |
| 55742 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction