missing translation for 'onlineSavingsMsg'
Learn More

alpha-p-Xylenesulfonyl chloride, 97%

Product Code. 15493347
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493347 Supplier Thermo Scientific Alfa Aesar Supplier No. H26038.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51419-59-1 | C8H9ClO2S | 204.668 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51419-59-1
Melting Point 75°C to 81°C
Molecular Formula C8H9ClO2S
MDL Number MFCD01631931
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Synonym 4-methylbenzylsulfonyl chloride, 4-methylphenyl methanesulfonyl chloride, p-tolylmethanesulfonyl chloride, p-tolyl-methanesulfonyl chloride, p-tolyl methanesulfonyl chloride, 4-methylphenyl methyl sulfonyl chloride, pubchem5592, pubchem5631, acmc-209wgt, 4-methylbenzylsulfonylchloride
InChI Key JALKUHLLMWYIAT-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)CS(=O)(=O)Cl
IUPAC Name (4-methylphenyl)methanesulfonyl chloride
Molecular Weight (g/mol) 204.668
PubChem CID 2794648
Formula Weight 204.67
Percent Purity 97%
Chemical Name or Material alpha-p-Xylenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.