missing translation for 'onlineSavingsMsg'
Learn More

Alpha-(p-Toluenesulfonyl)-4-fluorobenzylisonitrile, TRC

Product Code. 30423457
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423457 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423457 Supplier Toronto Research Chemicals Supplier No. TRCP768818100MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 165806-95-1
Molecular Formula C15H12FNO2S
Quantity 100 mg
InChI Formula InChI=1S/C15H12FNO2S/c1-11-3-9-14(10-4-11)20(18,19)15(17-2)12-5-7-13(16)8-6-12/h3-10,15H,1H3
SMILES Cc1ccc(cc1)S(=O)(=O)C([N+]#[C-])c2ccc(F)cc2
IUPAC Name 1-[(4-fluorophenyl)-isocyanomethyl]sulfonyl-4-methylbenzene
Molecular Weight (g/mol) 289.33
Formula Weight 289.0573
Chemical Name or Material alpha-(p-Toluenesulfonyl)-4-fluorobenzylisonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.