missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-N-terminal Methyltransferase 1A/METTL11A Antibody [FITC], Novus Biologicals™
Shop All Bio Techne ProductsDescription
alpha-N-terminal Methyltransferase 1A/METTL11A Polyclonal antibody specifically detects alpha-N-terminal Methyltransferase 1A/METTL11A in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | alpha-N-terminal Methyltransferase 1A/METTL11A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | AD-003, alpha N-terminal protein methyltransferase 1A, C9orf32, chromosome 9 open reading frame 32, EC 2.1.1.-, methyltransferase like 11A, Methyltransferase-like protein 11A, NRMT, N-terminal RCC1 methyltransferase, NTM1A, NTMT1, X-Pro-Lys N-terminal protein methyltransferase 1A |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant human alpha -N-terminal Methyltransferase 1A/METTL11A, Thr2-Arg223, Accession # Q9BV86 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?