missing translation for 'onlineSavingsMsg'
Learn More

alpha-N-terminal Methyltransferase 1A/METTL11A Antibody [DyLight 650], Novus Biologicals™

Product Code. 30737743 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737743 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737743 Supplier Novus Biologicals Supplier No. AF7159W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

alpha-N-terminal Methyltransferase 1A/METTL11A Polyclonal antibody specifically detects alpha-N-terminal Methyltransferase 1A/METTL11A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-N-terminal Methyltransferase 1A/METTL11A
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias AD-003, alpha N-terminal protein methyltransferase 1A, C9orf32, chromosome 9 open reading frame 32, EC 2.1.1.-, methyltransferase like 11A, Methyltransferase-like protein 11A, NRMT, N-terminal RCC1 methyltransferase, NTM1A, NTMT1, X-Pro-Lys N-terminal protein methyltransferase 1A
Host Species Sheep
Immunogen E. coli-derived recombinant human alpha -N-terminal Methyltransferase 1A/METTL11A, Thr2-Arg223, Accession # Q9BV86
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 28989
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.