missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-N-acetylglucosaminidase/NAGLU Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
alpha-N-acetylglucosaminidase/NAGLU Polyclonal specifically detects alpha-N-acetylglucosaminidase/NAGLU in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | alpha-N-acetylglucosaminidase/NAGLU |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | EC 3.2.1.50, MPS3B, MPS-IIIB, N-acetyl-alpha-glucosaminidase, N-acetylglucosaminidase, alpha, NAGalpha-N-acetylglucosaminidase, UFHSD, UFHSD1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human alpha-N-acetylglucosaminidase/NAGLU (NP_000254.2). Peptide sequence LEALVDSVAQGIPFQQHQFDKNVFQLEQAFVLSKQRYPSQPRGDTVDLAK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?