missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-N-acetylgalactosaminidase/NAGA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00180-100ul
This item is not returnable.
View return policy
Description
alpha-N-acetylgalactosaminidase/NAGA Polyclonal antibody specifically detects alpha-N-acetylgalactosaminidase/NAGA in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| alpha-N-acetylgalactosaminidase/NAGA | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| alpha-N- (alpha-galactosidase B), alpha-N-acetylgalactosaminidase, EC 3.2.1, EC 3.2.1.49, GALB, N-acetylgalactosaminidase, alpha- | |
| Produced in rabbits immunized with purified, recombinant Human alpha-N-acetylgalactosaminidase/NAGA (Accession#: NP_000253.1; Met1-Gln411). | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 4668 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction