missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylcinnamaldehyde, predominantly (E), 97%

Product Code. 11498633
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498633 100 g 100g
11408643 500 g 500g
11418643 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498633 Supplier Thermo Scientific Alfa Aesar Supplier No. A10429.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-39-3 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-39-3
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00006976
InChI Key VLUMOWNVWOXZAU-CLFYSBASSA-N
Synonym alpha-methylcinnamaldehyde, a-methylcinnamaldehyde, 2-propenal, 2-methyl-3-phenyl, 2-methyl-3-phenylprop-2-enal, e-2-methyl-3-phenylpropenal, 2-methyl-3-phenylacrylaldehyde, e-2-methyl-3-phenylacrylaldehyde, 2-methyl-3-phenyl-2-propenal, 2-methyl-3-phenylacrolein, alpha-methyl-trans-cinnamaldehyde
PubChem CID 5372813
SMILES C\C(C=O)=C\C1=CC=CC=C1

Specifications

Density 1.05
Boiling Point 138°C to 140°C (14 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.605
Quantity 100 g
Beilstein 507514
Sensitivity Air Sensitive
IUPAC Name (E)-2-methyl-3-phenylprop-2-enal
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material alpha-Methylcinnamaldehyde, predominantly ( E)
Show More Show Less
compliance-icons
Product Identifier
  • alpha-Methylcinnamaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: alpha-Methylcinnamaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.