missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Methylbenzenemethanol, TRC

Product Code. 30005044
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005044 100 g 100g
30004812 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005044 Supplier Toronto Research Chemicals Supplier No. TRCM226185100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-85-1
Molecular Formula C8 H10 O
Molecular Weight (g/mol) 122.16
Synonym Benzenemethanol, α-methyl- (9CI, ACI), Benzyl alcohol, α-methyl- (8CI), α-Methylbenzenemethanol (ACI), (1-Hydroxyethyl)benzene, (RS)-1-Phenylethanol, (RS)-sec-Phenethyl alcohol, (RS)-α-Phenylethanol, (±)-1-Phenylethanol, (±)-1-Phenylethyl alcohol, (±)-sec-Phenethyl alcohol, (±)-sec-Phenylethanol, (±)-α-Methylbenzenemethanol, (±)-α-Methylbenzyl alcohol, (±)-α-Phenethyl alcohol, (±)-α-Phenylethanol, 1-Hydroxy-1-phenylethane, 1-Phenethyl alcohol, 1-Phenyl-1-ethanol, 1-Phenyl-1-hydroxyethane, 1-Phenylethane-1-ol, 1-Phenylethanol, 1-Phenylethyl alcohol, DL-Methyl phenyl carbinol, DL-α-Phenylethyl alcohol, dl-1-Phenylethanol, Ethanol, 1-phenyl-, Methylphenylcarbinol, NSC 25502, Phenylmethylcarbinol, sec-Phenethyl alcohol, Styrallyl alcohol, Styralyl alcohol, Synthacolin, α-Hydroxyethylbenzene, α-Methylbenzyl alcohol, α-Methylphenylmethanol, α-Phenethyl alcohol, α-Phenylethanol, α-Phenylethyl alcohol
IUPAC Name 1-phenylethanol
SMILES CC(O)c1ccccc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3
Formula Weight 122.0732
Chemical Name or Material 1-Phenylethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.