missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Methylacyl-CoA Racemase/AMACR Antibody (CL9362) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88931MFV610
This item is not returnable.
View return policy
Description
alpha-Methylacyl-CoA Racemase/AMACR Monoclonal antibody specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| alpha-Methylacyl-CoA Racemase/AMACR | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM | |
| This antibody was developed using a recombinant protein derived from Q9UHK6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
| 23600 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL9362 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction