missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-Melanocyte Stimulating Hormone Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43691
This item is not returnable.
View return policy
Description
Alpha-Melanocyte Stimulating Hormone Polyclonal antibody specifically detects Alpha-Melanocyte Stimulating Hormone in Bovine,Human,Mouse,Porcine,Rabbit,Rat,Sheep samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Alpha-Melanocyte Stimulating Hormone | |
| Polyclonal | |
| Unconjugated | |
| PBS, pH 7.4, 50% glycerol | |
| Immunized with OVA - Alpha-Melanocyte Stimulating Hormone (Ser138∽Val150) | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 0.5-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Bovine, Human, Mouse, Pig, Rabbit, Rat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction