missing translation for 'onlineSavingsMsg'
Learn More

Alpha-mannosidase II Antibody (1G9), Novus Biologicals™

Product Code. 18338897 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338897 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338897 Supplier Novus Biologicals Supplier No. H00004124M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Alpha-mannosidase II Monoclonal antibody specifically detects Alpha-mannosidase II in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha-mannosidase II
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1G9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002363
Gene Alias alpha-mannosidase 2, AMan II, EC 3.2.1, EC 3.2.1.114, Golgi alpha-mannosidase II, golgi integral membrane protein 7, GOLIM7, Man II, MANA2MANII, Mannosidase alpha class 2A member 1, mannosidase, alpha type II, mannosidase, alpha, class 2A, member 1, Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase
Host Species Mouse
Immunogen MAN2A1 (NP_002363, 1045 a.a. ~ 1144 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DIHLVNLRTIQSKVGNGHSNEAALILHRKGFDCRFSSKGTGLFCSTTQGKILVQKLLNKFIVESLTPSSLSLMHSPPGTQNISEINLSPMEISTFRIQLR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4124
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.