missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alpha-lactalbumin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122245
This item is not returnable.
View return policy
Description
Alpha-lactalbumin Polyclonal Antibody for ICC/IF, IHC, Western Blot
This gene encodes alpha-lactalbumin, a principal protein of milk. Alpha-lactalbumin forms the regulatory subunit of the lactose synthase (LS) heterodimer and beta 1,4-galactosyltransferase (beta4Gal-T1) forms the catalytic component. Together, these proteins enable LS to produce lactose by transfering galactose moieties to glucose. As a monomer, alpha-lactalbumin strongly binds calcium and zinc ions and may possess bactericidal or antitumor activity. A folding variant of alpha-lactalbumin, called HAMLET, likely induces apoptosis in tumor and immature cells.
Specifications
| Alpha-lactalbumin | |
| Polyclonal | |
| Unconjugated | |
| LALBA | |
| ALACTA; a-LACTA; alfaLA; alpha lactabumin; alpha lactalbumin; alpha-lactalbumin; alpha-lactalbumin A; alpha-lactalbumin B; alpha-lactalbumin precursor (EC 2.4.1.22); alpha-lactalbumin precursor protein; alpha-lactalbumin protein variant D; Bos d 4; lactalbumin; lactalbumin alpha; lactalbumin, alpha; lactalbumin, alpha-; lactalbumin, alpha- precursor; lactose synthase B protein; Lalba; lysozyme G; lysozyme-like protein 7; LYZG; LYZL7; MGC138521; MGC138523 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 16770, 281894, 3906 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P00709, P00711, P29752 | |
| LALBA | |
| Recombinant protein Alpha-Lactalbumin. The antigen corresponds to amino acid range 24-141 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction