missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ketoglutaric acid, Free Acid, >98%, Thermo Scientific™

Product Code. 15835168 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15835168 100 g Plastic bottle 100g
15845168 500 g Plastic bottle 500g
15835308 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15835168 Supplier Thermo Scientific Supplier No. J17950.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-50-7
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.098
MDL Number MFCD00004165
InChI Key KPGXRSRHYNQIFN-UHFFFAOYSA-N
Synonym 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
PubChem CID 51
ChEBI CHEBI:30915
IUPAC Name 2-oxopentanedioic acid
SMILES C(CC(=O)O)C(=O)C(=O)O

Specifications

Melting Point 113°C to 117°C
Packaging Plastic bottle
Quantity 100 g
Beilstein 1705689
Formula Weight 146.1
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material alpha-Ketoglutaric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.