missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Keto Isovaleric Acid Sodium Salt, TRC

Product Code. 30012681
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012681 100 mg 100mg
30006265 1 g 1g
30008145 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012681 Supplier Toronto Research Chemicals Supplier No. TRCK193500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3715-29-5
Molecular Formula C5 H7 O3 . Na
Molecular Weight (g/mol) 138.1
Synonym Butanoic acid, 3-methyl-2-oxo-, sodium salt (1:1) (ACI), Butanoic acid, 3-methyl-2-oxo-, sodium salt (9CI), Butyric acid, 3-methyl-2-oxo-, sodium salt (8CI), 3-Methyl-2-oxobutanoic acid sodium salt, Sodium 3-methyl-2-oxobutanoate, Sodium 3-methyl-2-oxobutyrate, Sodium dimethylpyruvate, Sodium α-ketoisovalerate, Sodium α-oxoisovalerate
IUPAC Name sodium;3-methyl-2-oxobutanoate
SMILES [Na+].CC(C)C(=O)C(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1
Formula Weight 138.0293
Chemical Name or Material alpha-Keto Isovaleric Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.