missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Alpha-Kainic Acid Lithium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Alpha-Kainic Acid Lithium Salt |
| Analyte or Component Names | (-)-Alpha-Kainic Acid Lithium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H14 Li N O4 |
| Synonym | Lithium 2-((2S,3S,4S)-2-carboxy-4-(prop-1-en-2-yl)pyrrolidin-3-yl)acetate |
| InChI Formula | InChI=1S/C10H15NO4.Li/c1-5(2)7-4-11-9(10(14)15)6(7)3-8(12)13;/h6-7,9,11H,1,3-4H2,2H3,(H,12,13)(H,14,15);/q;+1/p-1/t6-,7+,9-;/m0./s1 |
| Solution Type | Neat |
| SMILES | [Li]OC(=O)C[C@H]1[C@H](CN[C@@H]1C(=O)O)C(=C)C |
| IUPAC Name | [2-[(2S,3S,4S)-2-carboxy-4-prop-1-en-2-ylpyrrolidin-3-yl]acetyl]oxylithium |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?