missing translation for 'onlineSavingsMsg'
Learn More

alpha-Isopropylphenylacetic acid, 97%

Product Code. 11473087
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473087 25 g 25g
11463087 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473087 Supplier Thermo Scientific Alfa Aesar Supplier No. A16077.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3508-94-9 | C11H14O2 | 178.231 g/mol

alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3508-94-9
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00021708
InChI Key HDLQGISFYDYWFJ-UHFFFAOYSA-N
Synonym alpha-isopropylphenylacetic acid, 2-isopropyl-2-phenylacetic acid, 3-methyl-2-phenyl-butyric acid, 3-methyl-2-phenylbutyric acid, butyric acid, 3-methyl-2-phenyl, benzeneacetic acid, alpha-1-methylethyl, 2-phenylisovaleric acid, acmc-1bn3m, benzeneacetic acid, alpha-1-methylethyl-9ci, alpha-isopropylphenylaceticacid
PubChem CID 19024
IUPAC Name 3-methyl-2-phenylbutanoic acid
SMILES CC(C)C(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 57°C to 62°C
Quantity 25 g
Beilstein 2045915
Solubility Information Slightly soluble in water.
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material alpha-Isopropylphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.