missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Internexin Recombinant Rabbit Monoclonal Antibody (23GB6325), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | alpha Internexin |
|---|---|
| Clone | 23GB6325 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089477
![]() |
Invitrogen™
MA553022 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087844
|
Invitrogen™
MA553023 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha Internexin Recombinant Monoclonal antibody specifically detects alpha Internexin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| alpha Internexin | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P23565, P46660, Q16352 | |
| 226180, 24503, 9118 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6325 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 | |
| A synthesized peptide derived from human alpha Internexin (400-499AA). | |
| Primary | |
| -20°C | |
| INA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title