missing translation for 'onlineSavingsMsg'
Learn More

alpha-Internexin Rabbit anti-Human, Mouse, Rat, Clone: 5E5Z3, Novus Biologicals™

Product Code. 18395224
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395224 100 μg 100µL
18350722 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18395224 Supplier Novus Biologicals Supplier No. NBP316230100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

alpha-Internexin Monoclonal antibody specifically detects alpha-Internexin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Internexin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5E5Z3
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human alpha-Internexin (Q16352).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9118
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.