missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Internexin Antibody (S03-6H1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19567-100ul
This item is not returnable.
View return policy
Description
alpha-Internexin Monoclonal antibody specifically detects alpha-Internexin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| alpha-Internexin | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-6H1 | |
| Unconjugated | |
| 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1 | |
| A synthetic peptide of human alpha-Internexin (Uniprot # Q16352) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 9118 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction