missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Internexin Antibody (2E3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-140H
This item is not returnable.
View return policy
Description
alpha-Internexin Monoclonal antibody specifically detects alpha-Internexin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| alpha-Internexin | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2000 | |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1 | |
| Purified recombinant rat alpha Internexin expressed in and purified from E. coli [UniProt# P23565] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 9118 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction