missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Hyodeoxycholic Acid Methyl Ester, TRC

Product Code. 30394986
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394986 10 g -
30408631 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394986 Supplier Toronto Research Chemicals Supplier No. TRCH99811010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2868-48-6
Molecular Formula C25H42O4
Molecular Weight (g/mol) 406.6
Synonym Hyodeoxycholic Acid Methyl Ester, Methyl 3alpha,6alpha-Dihydroxy-5beta-cholan-24-oate, Methyl 3alpha,6alpha-Dihydroxy-5beta-cholanate, Methyl 3alpha,6alpha-dihydroxy-5beta-cholanoate, Methyl Hyodeoxycholanate, Methyl Hyodeoxycholate, alpha-Hyodeoxycholic Acid Methyl Ester
IUPAC Name methyl (4R)-4-[(3R,5R,6S,8S,9S,10R,13R,14S,17R)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3C[C@H](O)[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C25H42O4/c1-15(5-8-23(28)29-4)18-6-7-19-17-14-22(27)21-13-16(26)9-11-25(21,3)20(17)10-12-24(18,19)2/h15-22,26-27H,5-14H2,1-4H3/t15-,16-,17+,18-,19+,20+,21+,22+,24-,25-/m1/s1
Formula Weight 406.3083
Chemical Name or Material alpha-Hyodeoxycholic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.