missing translation for 'onlineSavingsMsg'
Learn More

alpha-Hydroxymethyl Atropine, TRC

Product Code. 30393458
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393458 50 mg
30399099 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393458 Supplier Toronto Research Chemicals Supplier No. TRCH94640050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2515-36-8
Molecular Formula C18 H25 N O4
Molecular Weight (g/mol) 319.4
Synonym α-Hydroxymethyl Atropine, Hydracrylic acid, 2-(hydroxymethyl)-2-phenyl-, 3α-tropanyl ester (7CI), alpha-Hydroxymethylatropine
IUPAC Name [(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-(hydroxymethyl)-2-phenylpropanoate
SMILES CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)C(CO)(CO)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H25NO4/c1-19-14-7-8-15(19)10-16(9-14)23-17(22)18(11-20,12-21)13-5-3-2-4-6-13/h2-6,14-16,20-21H,7-12H2,1H3/t14-,15+,16+
Formula Weight 319.1784
Chemical Name or Material alpha-Hydroxymethyl Atropine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.