missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-Hydroxy Montelukast, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-Hydroxy Montelukast |
| CAS | 2045402-27-3 |
| Molecular Formula | C35 H36 Cl N O4 S |
| Quantity | 10 mg |
| Synonym | Montelukast Sodium Imp. I (EP), Cyclopropaneacetic acid, 1-[[[(1R)-1-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-α-hydroxy- |
| InChI Formula | InChI=1S/C35H36ClNO4S/c1-34(2,41)29-9-4-3-7-24(29)13-17-31(42-22-35(18-19-35)32(38)33(39)40)26-8-5-6-23(20-26)10-15-28-16-12-25-11-14-27(36)21-30(25)37-28/h3-12,14-16,20-21,31-32,38,41H,13,17-19,22H2,1-2H3,(H,39,40)/b15-10+/t31-,32?/m1/s1 |
| SMILES | CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC2)C(O)C(=O)O)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3 |
| IUPAC Name | 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]-2-hydroxyacetic acid |
| Molecular Weight (g/mol) | 602.18 |
| Formula Weight | 601.2054 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?