missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Hederin, TRC

Product Code. 17991255
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991255 50 mg 50mg
17994505 5 mg 5mg
17931795 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991255 Supplier Toronto Research Chemicals Supplier No. TRCH12999550MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27013-91-8
Molecular Formula C41 H66 O12
Molecular Weight (g/mol) 750.96
Synonym Olean-12-en-28-oic acid, 3-[[2-O-(6-deoxy-α-L-mannopyranosyl)-α-L-arabinopyranosyl]oxy]-23-hydroxy-, (3β,4α)-, α-Hederin (7CI,8CI), (+)-Dipsacobioside, (3β,4α)-3-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-α-L-arabinopyranosyl]oxy]-23-hydroxyolean-12-en-28-oic acid, 23-Hydroxy-3β-[(O-α-L-rhamnopyranosyl-(1→2)-α-L-arabinopyranosyl)oxy]olean-12-en-28-oic acid, 3-O-α-L-Rhamnopyranosyl-(1→2)-α-L-arabinopyranosylhederagenin, Akebia saponin PD, Akeboside Stc, Cephalaroside C, Dipsacobioside, Dipsacoside A, Dipsacus saponin L, Glycoside L-E1, Hederagenin 3-O-α-L-rhamnopyranosyl-(1 → 2)-α-L-arabinopyranoside, Hederoside C, Helixin, Kalopanaxsaponin A, Kizuta saponin K6, Koronaroside A, NSC 106553, Nepalin 2, Prosapogenin CP3b, Pulsatilla saponin A, Sapindoside A, Tauroside E
IUPAC Name (4aS,6aR,6aS,6bR,8aR,9R,10S,12aR,14bS)-10-[(2S,3R,4S,5S)-4,5-dihydroxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-9-(hydroxymethyl)-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)CO[C@H]2O[C@H]3CC[C@@]4(C)[C@@H](CC[C@]5(C)[C@@H]4CC=C6[C@@H]7CC(C)(C)CC[C@@]7(CC[C@@]56C)C(=O)O)[C@]3(C)CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C41H66O12/c1-21-28(44)30(46)31(47)33(51-21)53-32-29(45)24(43)19-50-34(32)52-27-11-12-37(4)25(38(27,5)20-42)10-13-40(7)26(37)9-8-22-23-18-36(2,3)14-16-41(23,35(48)49)17-15-39(22,40)6/h8,21,23-34,42-47H,9-20H2,1-7H3,(H,48,49)/t21-,23-,24-,25+,26+,27-,28-,29-,30+,31+,32+,33-,34-,37-,38-,39+,40+,41-/m0/s1
Formula Weight 750.4554
Percent Purity >95%
Chemical Name or Material alpha-Hederin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.