missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-Glycerophosphoric Acid-13C3 Bis-cyclohexylammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Alpha-Glycerophosphoric Acid-13C3 Bis-cyclohexylammonium Salt |
| Molecular Formula | C1213C3H35N2O6P |
| Quantity | 1 mg |
| Synonym | 1,2,3-Propanetriol-13C3 1-(Dihydrogen Phosphate) Cyclohexanamine, Sα-Glycerophosphate-13C3 Cyclohexylamine Salt, Glycerol-13C3 3-Phosphate Dicyclohexylammonium Salt |
| InChI Formula | InChI=1S/2C6H13N.C3H9O6P/c2*7-6-4-2-1-3-5-6;4-1-3(5)2-9-10(6,7)8/h2*6H,1-5,7H2;3-5H,1-2H2,(H2,6,7,8)/i;;1+1,2+1,3+1 |
| SMILES | NC1CCCCC1.NC2CCCCC2.O[13CH2][13CH](O)[13CH2]OP(=O)(O)O |
| Molecular Weight (g/mol) | 373.4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?