missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
alpha Glucosidase 2 Polyclonal antibody specifically detects alpha Glucosidase 2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | alpha Glucosidase 2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_938148.1 |
| Gene Alias | Alpha-glucosidase 2, EC 3.2.1, EC 3.2.1.84, G2ANGLUII, Glucosidase II subunit alpha, glucosidase, alpha, neutral AB, GluII, KIAA0088alpha glucosidase II alpha subunit, neutral alpha-glucosidase AB |
| Host Species | Rabbit |
| Immunogen | GANAB (NP_938148.1, 1 a.a. - 944 a.a.) full-length human protein. MAAVAAVAARRRRSWASLVLAFLGVCLGITLAVDRSNFKTCEESSFCKRQRSIRPGLSPYRALLDSLQLGPDSLTVHLIHEVTKVLLVLELQGLQKNMTRFRIDELEPRRPRYRVPDVLVADPPIARLSVSGRDENSVELTMAEGPYKIILTARPFRLDLLEDRSLLLSVNARGLLEFEHQRAPRVSQGSKDPAEGDGAQPEETPRDGDKPEETQGKAEKDEPGAWEETFKTHSDSKPYGPMSVGLDFSLPGMEHVYGIPEHADNLRLKVTEGGEPYRLYNLDVFQYELYNPMALYGSVPVLLAHNPHRDLGIFWLNAAETWVDISSNTAGKTLFGKMMDYLQGSGETPQTDVRWMSETGIIDVFLLLGPSISDVFRQYASLTGTQALPPLFSLGYHQSRWNYRDEADVLEVDQGFDDHNLPCDVIWLDIEHADGKRYFTWDPSRFPQPRTMLERLASKRRKLVAIVDPHIKVDSGYRVHEELRNLGLYVKTRDGSDYEGWCWPGSAGYPDFTNPTMRAWWANMFSYDNYEGSAPNLFVWNDMNEPSVFNGPEVTMLKDAQHYGGWEHRDVHNIYGLYVHMATADGLRQRSGGMERPFVLARAFFAGSQRFGAVWTGDNTAEWDHLKISIPMCLSLGLVGLSFCGADVGGFFKNPEPELLVRWYQMGAYQPFFRAHAHLDTGRREPWLLPSQHNDIIRDALGQRYSLLPFWYTLLYQAHREGIPVMRPLWVQYPQDVTTFNIDDQYLLGDALLVHPVSDSGAHGVQVYLPGQGEVWYDIQSYQKHHGPQTLYLPVTLSSIPVFQRGGTIVPRWMRVRRSSECMKDDPITLFVALSPQGTAQGELFLDDGHTFNYQTRQEFLLRRFSFSGNTLVSSSADPEGHFETPIWIERVVIIGAGKPAAVVLQTKGSPESRLSFQHDPETSVLVLRKPGINVASDWSIHLR |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?